2-Methyl-2H-indazol-4-amine ≥98%

Supplier: Matrix Scientific

Synonyms: 4-Amino-2-methyl-2H-indazole

069655-500MG 069655-1G 069655-5G
101941-288EA 553.85 USD
101941-288 101941-290 101941-292
2-Methyl-2H-indazol-4-amine ≥98%
2-Methyl-2H-indazol-4-amine

Hazard: Irritant

Formula: C₈H₉N₃
MW: 147.1800 g/mol
MDL Number: MFCD09870044
CAS Number: 82013-51-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR