1-Methyl-1H-indazol-7-ylamine ≥97%

Supplier: Matrix Scientific

Synonyms: 7-Amino-1-methylindazole, 7-Amino-1-methyl-1H-indazole

069654-1G 069654-5G
101941-284EA 200 USD
101941-284 101941-286
1-Methyl-1H-indazol-7-ylamine ≥97%
1-Methyl-1H-indazol-7-ylamine

Hazard: Irritant

Formula: C₈H₉N₃
MDL Number: MFCD09870043
CAS Number: 41926-06-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR