4-Bromo-1-methyl-1H-indazole ≥98%
Supplier: Matrix Scientific
069653-1G
069653-5G
101941-280EA
263.08
USD
101941-280
101941-282
4-Bromo-1-methyl-1H-indazole ≥98%
4-Bromo-1-methyl-1H-indazole
Hazard: Irritant
Formula:
C₈H₇BrN₂ MW: 211.0586 g/mol |
MDL Number:
MFCD09870042 CAS Number: 365427-30-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...