2-(N-Anilino)pyrimidine ≥98%

Supplier: Matrix Scientific

Synonyms: N-Phenylpyrimidin-2-amine

069648-1G 069648-5G 069648-25G
101941-256EA 233.85 USD
101941-256 101941-258 101941-260
2-(N-Anilino)pyrimidine ≥98%
2-(N-Anilino)pyrimidine

Hazard: Irritant

Formula: C₁₀H₉N₃
MW: 171.2 g/mol
MDL Number: MFCD01646270
CAS Number: 57356-49-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR