3,4-Diisopropoxy-3-cyclobutene-1,2-dione ≥99%

Supplier: Matrix Scientific

Synonyms: Diisopropyl Squarate, 3,4-Diisopropoxycyclobut-3-ene-1,2-dione

069639-1G 069639-5G
101941-214EA 70.77 USD
101941-214 101941-216
3,4-Diisopropoxy-3-cyclobutene-1,2-dione ≥99%
3,4-Diisopropoxy-3-cyclobutene-1,2-dione

Hazard: Irritant

Formula: C₁₀H₁₄O₄
MW: 198.22 g/mol
MDL Number: MFCD00075125
CAS Number: 61699-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR