2-Chloro-6-trifluoromethyl-3-(trimethylsilyl)pyridine ≥97%

Supplier: Matrix Scientific

069637-1G 069637-5G
101941-206EA 320 USD
101941-206 101941-208
2-Chloro-6-trifluoromethyl-3-(trimethylsilyl)pyridine ≥97%
2-Chloro-6-trifluoromethyl-3-(trimethylsilyl)pyridine

Hazard: Irritant

Formula: C₉H₁₁ClF₃NSi
MDL Number: MFCD09763705
CAS Number: 205444-18-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR