2-Chloro-3-(trimethylsilyl)pyridine ≥96%

Supplier: Matrix Scientific

069636-1G 069636-5G 069636-25G
101941-200EA 233.85 USD
101941-200 101941-202 101941-204
2-Chloro-3-(trimethylsilyl)pyridine ≥96%
2-Chloro-3-(trimethylsilyl)pyridine

Hazard: Irritant

Formula: C₈H₁₂ClNSi
MDL Number: MFCD09763704
CAS Number: 77332-76-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR