(4-Methyl-1H-imidazol-5-yl)methanol hydrochloride ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Hydroxymethyl-5-methylimidazole hydrochloride

069635-5G 069635-25G
101941-196EA 18.46 USD
101941-196 101941-198
(4-Methyl-1H-imidazol-5-yl)methanol hydrochloride ≥97%
(4-Methyl-1H-imidazol-5-yl)methanol hydrochloride

Hazard: Irritant

Formula: C₅H₉ClN₂O
MW: 148.5908 g/mol
MDL Number: MFCD00040986
CAS Number: 38585-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR