4-N-(Boc)aminomethylphenylboronic acid ≥97%

Supplier: Matrix Scientific

Synonyms: (4-(((tert-Butoxycarbonyl)amino)methyl)phenyl)boronic acid, 4-(tert-Butoxycarbonylaminomethyl)phenylboronic acid, 4-(Aminomethyl)benzeneboronic acid, N-BOC protected

069632-1G 069632-5G
101941-182EA 349.23 USD
101941-182 101941-184
4-N-(Boc)aminomethylphenylboronic acid ≥97%
4-N-(Boc)aminomethylphenylboronic acid

Hazard: Irritant

Formula: C₁₂H₁₈BNO₄
MW: 251.09 g/mol
MDL Number: MFCD04115637
CAS Number: 489446-42-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR