4-Amino-2-chloro-5-fluoropyrimidine ≥98%
Supplier: Matrix Scientific
069631-1G
069631-5G
069631-25G
101941-176EA
64.62
USD
101941-176
101941-178
101941-180
4-Amino-2-chloro-5-fluoropyrimidine ≥98%
4-Amino-2-chloro-5-fluoropyrimidine
Hazard: Irritant
Formula:
C₄H₃ClFN₃ MW: 147.54 g/mol |
MDL Number:
MFCD00057344 CAS Number: 155-10-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...