Dimethyl-2-bromo-1H-imidazole-4,5-dicarboxylate ≥98%
Supplier: Matrix Scientific
Synonyms:
Dimethyl 2-bromo-1H-imidazole-4,5-dicarboxylate
069630-1G
069630-5G
101941-172EA
524.62
USD
101941-172
101941-174
Dimethyl-2-bromo-1H-imidazole-4,5-dicarboxylate ≥98%
Dimethyl-2-bromo-1H-imidazole-4,5-dicarboxylate
Hazard: Irritant
Formula:
C₇H₇BrN₂O₄ MW: 263.05 g/mol |
MDL Number:
MFCD09702026 CAS Number: 705280-65-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...