5-Bromo-4-N,N-dimethylamino-2-methoxypyrimidine ≥98%

Supplier: Matrix Scientific

069629-1G 069629-5G 069629-25G
101941-166EA 116.92 USD
101941-166 101941-168 101941-170
5-Bromo-4-N,N-dimethylamino-2-methoxypyrimidine ≥98%
5-Bromo-4-N,N-dimethylamino-2-methoxypyrimidine

Hazard: Irritant

Formula: C₇H₁₀BrN₃O
MW: 232.09 g/mol
MDL Number: MFCD07781216
CAS Number: 57054-81-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR