tert-Butyl-5-bromo-2-(methylthio)-6-oxopyrimidine-1-acetate ≥97%

Supplier: Matrix Scientific

069628-500MG 069628-1G 069628-5G
101941-160EA 640 USD
101941-160 101941-162 101941-164
tert-Butyl-5-bromo-2-(methylthio)-6-oxopyrimidine-1-acetate ≥97%
tert-Butyl-5-bromo-2-(methylthio)-6-oxopyrimidine-1-acetate

Hazard: Irritant

Formula: C₁₁H₁₅BrN₂O₃S
MDL Number: MFCD09702025
CAS Number: 308276-66-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR