2-Methyl-1-(tributylstannyl)-1-propene ≥96%

Supplier: Matrix Scientific

069627-1G 069627-5G
101941-156EA 372.31 USD
101941-156 101941-158
2-Methyl-1-(tributylstannyl)-1-propene ≥96%
2-Methyl-1-(tributylstannyl)-1-propene

Hazard: Irritant

Formula: C₁₆H₃₄Sn
MDL Number: MFCD09750019
CAS Number: 66680-86-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR