5-Bromo-2-chloro-4-(dimethylamino)pyrimidine ≥98%

Supplier: Matrix Scientific

069625-1G 069625-5G 069625-25G
101941-144EA 64.62 USD
101941-144 101941-146 101941-148
5-Bromo-2-chloro-4-(dimethylamino)pyrimidine ≥98%
5-Bromo-2-chloro-4-(dimethylamino)pyrimidine

Hazard: Irritant

Formula: C₆H₇BrClN₃
CAS Number: 57054-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR