2-Chloro-4-(hydroxymethyl)thiazole ≥97%

Supplier: Matrix Scientific

069623-500MG 069623-1G 069623-5G
101941-132EA 246.15 USD
101941-132 101941-134 101941-136
2-Chloro-4-(hydroxymethyl)thiazole ≥97%
2-Chloro-4-(hydroxymethyl)thiazole

Hazard: Irritant

Formula: C₄H₄ClNOS
MDL Number: MFCD09265529
CAS Number: 5198-85-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR