Methyl-2-amino-5-oxazolecarboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 2-aminooxazole-5-carboxylate

069621-1G 069621-25G
101941-122EA 263.08 USD
101941-122 101941-126
Methyl-2-amino-5-oxazolecarboxylate ≥97%
Methyl 2-amino-5-oxazolecarboxylate

Hazard: Irritant

Formula: C₅H₆N₂O₃
MW: 142.112 g/mol
MDL Number: MFCD09265526
CAS Number: 934236-40-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR