2-Amino-5-bromo-4-methylthiazole hydrochloride ≥98%

Supplier: Matrix Scientific

069617-1G 069617-5G
101941-108EA 349.23 USD
101941-108 101941-110
2-Amino-5-bromo-4-methylthiazole hydrochloride ≥98%
2-Amino-5-bromo-4-methylthiazole hydrochloride

Hazard: Irritant

Formula: C₄H₆BrClN₂S
MDL Number: MFCD09265523
CAS Number: 133692-16-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR