2-Amino-5-chloro-4-methylthiazole ≥97%

Supplier: Matrix Scientific

069616-1G 069616-5G
101941-104EA 349.23 USD
101941-104 101941-106
2-Amino-5-chloro-4-methylthiazole ≥97%
2-Amino-5-chloro-4-methylthiazole

Hazard: Irritant

Formula: C₄H₅ClN₂S
MW: 148.61 g/mol
MDL Number: MFCD10699130
CAS Number: 5316-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR