2-Chloro-4-formylthiazole

Supplier: Matrix Scientific

069615-1G 069615-5G
101941-100EA 429.23 USD
101941-100 101941-102
2-Chloro-4-formylthiazole
2-Chloro-4-formylthiazole

Hazard: Irritant

Formula: C₄H₂ClNOS
MDL Number: MFCD09702023
CAS Number: 5198-79-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR