3-(tert-Butoxycarbonyl)phenylboronic acid pinacol ester ≥98%

Supplier: Matrix Scientific

Synonyms: 3-(t-Butoxycarbonyl)phenylboronic acid pinacol ester, tert-Butyl 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

069614-1G 069614-5G 069614-25G
101941-092EA 55.38 USD
101941-092 101941-094 101941-096
3-(tert-Butoxycarbonyl)phenylboronic acid pinacol ester ≥98%
3-(tert-Butoxycarbonyl)phenylboronic acid pinacol ester

Hazard: Irritant

Formula: C₁₇H₂₅BO₄
MW: 304.1890 g/mol
MDL Number: MFCD03453057
CAS Number: 903895-48-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR