Copper(I) diphenylphosphinate ≥90%

Supplier: Matrix Scientific

069613-1G 069613-25G 069613-100G
101941-086EA 44.62 USD
101941-086 101941-088 101941-090
Copper(I) diphenylphosphinate ≥90%
Copper(I) diphenylphosphinate

Hazard: Irritant

Formula: C₁₂H₁₀CuO₂P
MDL Number: MFCD09702021
CAS Number: 1011257-42-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR