Benzothiazole-2-sulfonyl fluoride ≥97%

Supplier: Matrix Scientific

069607-1G 069607-5G
101941-054EA 349.23 USD
101941-054 101941-056
Benzothiazole-2-sulfonyl fluoride ≥97%
Benzothiazole-2-sulfonyl fluoride

Hazard: Irritant

Formula: C₇H₄FNO₂S₂
MW: 217.24 g/mol
MDL Number: MFCD09265512
CAS Number: 878376-34-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR