Methyl-2-bromo-5-isopropylthiazole-4-carboxylate ≥95%

Supplier: Matrix Scientific

069601-1G 069601-5G 069601-25G
101941-020EA 349.23 USD
101941-020 101941-022 101941-024
Methyl-2-bromo-5-isopropylthiazole-4-carboxylate ≥95%
Methyl-2-bromo-5-isopropylthiazole-4-carboxylate

Hazard: Irritant

Formula: C₈H₁₀BrNO₂S
MW: 264.15 g/mol
MDL Number: MFCD09265507
CAS Number: 81569-28-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR