Methyl-2-chloro-5-isopropylthiazole-4-carboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 2-chloro-5-isopropylthiazole-4-carboxylate

069600-1G 069600-5G 069600-25G
101941-014EA 349.23 USD
101941-014 101941-016 101941-018
Methyl-2-chloro-5-isopropylthiazole-4-carboxylate ≥97%
Methyl-2-chloro-5-isopropylthiazole-4-carboxylate

Hazard: Irritant

Formula: C₈H₁₀ClNO₂S
MW: 219.69 g/mol
MDL Number: MFCD09265506
CAS Number: 81569-27-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR