2,3-Dihydrobenzofuran-5-boronic acid ≥98%
Supplier: Matrix Scientific
069599-1G
069599-5G
101941-008EA
436.92
USD
101941-008
101941-010
2,3-Dihydrobenzofuran-5-boronic acid ≥98%
2,3-Dihydrobenzofuran-5-boronic acid
Hazard: Irritant
Formula:
C₈H₉BO₃ MW: 163.97 g/mol |
MDL Number:
MFCD02681979 CAS Number: 227305-69-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...