Methyl-2-chlorooxazole-4-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-chlorooxazole-4-carboxylate

069598-1G 069598-5G
101941-004EA 121.54 USD
101941-004 101941-006
Methyl-2-chlorooxazole-4-carboxylate ≥95%
Methyl-2-chlorooxazole-4-carboxylate

Hazard: Irritant/Refrigerate

Formula: C₅H₄ClNO₃
MW: 161.55 g/mol
MDL Number: MFCD09265505
CAS Number: 934236-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR