Pyrimidine-2-sulfonyl fluoride ≥97%

Supplier: Matrix Scientific

069597-1G 069597-5G
101940-998EA 346.15 USD
101940-998 101941-000
Pyrimidine-2-sulfonyl fluoride ≥97%
Pyrimidine-2-sulfonyl fluoride

Hazard: Irritant

Formula: C₄H₃FN₂O₂S
MDL Number: MFCD09265503
CAS Number: 35762-87-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR