3'-Azido-3'-deoxythymidine ≥98%
Supplier: Matrix Scientific
Synonyms:
1-(3-Azido-2,3-dideoxy-beta-D-ribofuranosyl)thymine, Zidovudine, Azidothymidine, AZT
069595-250MG
069595-500MG
069595-1G
101940-988EA
466.15
USD
101940-988
101940-990
101940-992
3'-Azido-3'-deoxythymidine ≥98%
3'-Azido-3'-deoxythymidine
Hazard: Irritant
Formula:
C₁₀H₁₃N₅O₄ MW: 267.25 g/mol |
MDL Number:
MFCD00006536 CAS Number: 30516-87-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...