2-Chloro-5-(tributylstannyl)thiazole ≥95%
Supplier: Matrix Scientific
069593-1G
069593-5G
069593-25G
101940-976EA
481.54
USD
101940-976
101940-978
101940-980
2-Chloro-5-(tributylstannyl)thiazole ≥95%
2-Chloro-5-(tributylstannyl)thiazole
Hazard: Irritant
Formula:
C₁₅H₂₈ClNSSn |
MDL Number:
MFCD09265501 CAS Number: 889672-73-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...