Methyl-2-chlorothiazole-5-carboxylate ≥97%

Supplier: Matrix Scientific

069592-1G 069592-5G
101940-972EA 153.85 USD
101940-972 101940-974
Methyl-2-chlorothiazole-5-carboxylate ≥97%
Methyl-2-chlorothiazole-5-carboxylate

Hazard: Irritant

Formula: C₅H₄ClNO₂S
MDL Number: MFCD09265500
CAS Number: 72605-86-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR