6-Bromo-1-(phenylsulfonyl)-1H-indole ≥97%

Supplier: Matrix Scientific

069590-1G 069590-5G
101940-962EA 263.08 USD
101940-962 101940-964
6-Bromo-1-(phenylsulfonyl)-1H-indole ≥97%
6-Bromo-1-(phenylsulfonyl)-1H-indole

Hazard: Irritant

Formula: C₁₄H₁₀BrNO₂S
MW: 336.2000 g/mol
MDL Number: MFCD03840610
CAS Number: 679794-03-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR