3-Fluoro-2-(tributylstannyl)pyridine ≥96%

Supplier: Matrix Scientific

069585-1G 069585-5G
101940-938EA 290.77 USD
101940-938 101940-940
3-Fluoro-2-(tributylstannyl)pyridine ≥96%
3-Fluoro-2-(tributylstannyl)pyridine

Hazard: Irritant

Formula: C₁₇H₃₀FNSn
CAS Number: 573675-60-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR