4-Iodo-2-(methylthio)pyrimidine ≥96%

Supplier: Matrix Scientific

Synonyms: 4-Iodo-2-methylsulphanylpyrimidine

069583-1G 069583-5G 069583-25G
101940-928EA 233.85 USD
101940-928 101940-930 101940-932
4-Iodo-2-(methylthio)pyrimidine ≥96%
4-Iodo-2-(methylthio)pyrimidine

Hazard: Irritant

Formula: C₅H₅IN₂S
MW: 252.08 g/mol
MDL Number: MFCD01319020
CAS Number: 1122-74-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR