2-(N,N-Dimethylamino)-6-iodopyrazine ≥95%

Supplier: Matrix Scientific

069582-1G 069582-5G
101940-924EA 407.69 USD
101940-924 101940-926
2-(N,N-Dimethylamino)-6-iodopyrazine ≥95%
2-(N,N-Dimethylamino)-6-iodopyrazine

Hazard: Irritant

Formula: C₆H₈IN₃
MDL Number: MFCD09265493
CAS Number: 125060-66-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR