2-Amino-4-iodo-6-methoxypyrimidine ≥97%

Supplier: Matrix Scientific

069580-1G 069580-5G
101940-914EA 524.62 USD
101940-914 101940-916
2-Amino-4-iodo-6-methoxypyrimidine ≥97%
2-Amino-4-iodo-6-methoxypyrimidine

Hazard: Irritant

Formula: C₅H₆IN₃O
MDL Number: MFCD09265491
CAS Number: 100594-13-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR