Ethyl-4-bromo-5-methoxy-1H-indole-2-carboxylate

Supplier: Matrix Scientific

069577-250MG 069577-500MG 069577-1G
101940-896EA 349.23 USD
101940-896 101940-898 101940-900
Ethyl-4-bromo-5-methoxy-1H-indole-2-carboxylate
Ethyl-4-bromo-5-methoxy-1H-indole-2-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₂BrNO₃
MDL Number: MFCD09265488
CAS Number: 30933-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR