Ethyl-2-bromothiazole-5-carboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-bromothiazole-5-carboxylate

069576-5G 069576-25G
101940-892EA 61.54 USD
101940-892 101940-894
Ethyl-2-bromothiazole-5-carboxylate ≥97%
Ethyl-2-bromothiazole-5-carboxylate

Hazard: Irritant

Formula: C₆H₆BrNO₂S
MW: 236.08 g/mol
Boiling Pt: 216…217 °C
Density: 1.581
Flash Pt: 110 °C
MDL Number: MFCD00463837
CAS Number: 41731-83-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR