5-Bromo-2-methylsulfonylpyridine ≥98%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-2-(methylsulphonyl)pyridine

069575-1G 069575-5G
101940-886EA 246.15 USD
101940-886 101940-888
5-Bromo-2-methylsulfonylpyridine ≥98%
5-Bromo-2-methylsulfonylpyridine

Hazard: Irritant

Formula: C₆H₆BrNO₂S
MW: 236.09 g/mol
MDL Number: MFCD04974510
CAS Number: 98626-95-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR