5-Bromo-2-(methylthio)pyridine ≥97%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-2-methylthiopyridine, 2-Methylthio-5-bromopyridine

069574-1G 069574-5G 069574-25G
101940-880EA 263.08 USD
101940-880 101940-882 101940-884
5-Bromo-2-(methylthio)pyridine ≥97%
5-Bromo-2-(methylthio)pyridine

Hazard: Irritant

Formula: C₆H₆BrNS
MW: 204.0875 g/mol
MDL Number: MFCD03411571
CAS Number: 51933-78-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR