5-N-(Boc)-aminomethyl-3-isopropylisoxazole

Supplier: Matrix Scientific

069570-500MG 069570-1G 069570-5G
101940-858EA 349.23 USD
101940-858 101940-860 101940-862
5-N-(Boc)-aminomethyl-3-isopropylisoxazole
5-N-(Boc)-aminomethyl-3-isopropylisoxazole

Hazard: Irritant

Formula: C₁₂H₂₀N₂O₃
MDL Number: MFCD09265486
CAS Number: 543713-09-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR