Isobutyraldehyde oxime ≥95%

Supplier: Matrix Scientific

069569-1G 069569-5G 069569-25G
101940-852EA 92.31 USD
101940-852 101940-854 101940-856
Isobutyraldehyde oxime ≥95%
Isobutyraldehyde oxime

Hazard: Irritant

Formula: C₄H₉NO
MDL Number: MFCD00013940
CAS Number: 151-00-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR