2-Ethoxy-5-pyrimidineboronic acid ≥99%

Supplier: Matrix Scientific

Synonyms: (2-Ethoxypyrimidin-5-yl)boronic acid

069568-1G 069568-5G
101940-848EA 307.69 USD
101940-848 101940-850
2-Ethoxy-5-pyrimidineboronic acid ≥99%
2-Ethoxy-5-pyrimidineboronic acid

Hazard: Irritant

Formula: C₆H₉BN₂O₃
MW: 167.9600 g/mol
MDL Number: MFCD07375134
CAS Number: 1003043-55-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR