trans-Bis(tributylstannyl)ethylene ≥90%
Supplier: Matrix Scientific
Synonyms:
trans-1,2-Bis(tri-n-butylstannyl)ethylene
069567-1G
069567-5G
069567-25G
101940-842EA
160
USD
101940-842
101940-844
101940-846
trans-Bis(tributylstannyl)ethylene ≥90%
trans-Bis(tributylstannyl)ethylene
Hazard: Irritant/Refrigerate
Formula:
C₂₆H₅₆Sn₂ MW: 606.15 |
MDL Number:
MFCD01631299 CAS Number: 14275-61-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...