5-Bromo-2-(methylsulfonyl)pyrimidine ≥95%

Supplier: Matrix Scientific

069565-1G 069565-5G 069565-25G
101940-830EA 349.23 USD
101940-830 101940-832 101940-834
5-Bromo-2-(methylsulfonyl)pyrimidine ≥95%
5-Bromo-2-(methylsulfonyl)pyrimidine

Hazard: Irritant

Formula: C₅H₅BrN₂O₂S
MDL Number: MFCD00508247
CAS Number: 38275-48-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR