2-Methoxy-3-(tributylstannyl)pyrazine
Supplier: Matrix Scientific
069564-1G
069564-5G
101940-826EA
349.23
USD
101940-826
101940-828
2-Methoxy-3-(tributylstannyl)pyrazine
2-Methoxy-3-(tributylstannyl)pyrazine
Hazard: Irritant
Formula:
C₁₇H₃₂N₂OSn |
MDL Number:
MFCD09025812 CAS Number: 1105511-65-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...