4-Methyl-2-(tributylstannyl)thiazole ≥94%

Supplier: Matrix Scientific

069559-500MG 069559-1G 069559-5G
101940-802EA 355.38 USD
101940-802 101940-804 101940-806
4-Methyl-2-(tributylstannyl)thiazole ≥94%
4-Methyl-2-(tributylstannyl)thiazole

Hazard: Irritant

Formula: C₁₆H₃₁NSSn
MDL Number: MFCD09025808
CAS Number: 251635-59-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR