2-Ethoxy-5-(tributylstannyl)pyrimidine ≥95%

Supplier: Matrix Scientific

069558-500MG 069558-1G 069558-5G
101940-796EA 263.08 USD
101940-796 101940-798 101940-800
2-Ethoxy-5-(tributylstannyl)pyrimidine ≥95%
2-Ethoxy-5-(tributylstannyl)pyrimidine

Hazard: Irritant

Formula: C₁₈H₃₄N₂OSn
MDL Number: MFCD09025789
CAS Number: 1025746-10-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR