3-Methyl-2-(tributylstannyl)thiophene ≥94%

Supplier: Matrix Scientific

069557-500MG 069557-1G 069557-5G
101940-790EA 383.08 USD
101940-790 101940-792 101940-794
3-Methyl-2-(tributylstannyl)thiophene ≥94%
3-Methyl-2-(tributylstannyl)thiophene

Hazard: Irritant

Formula: C₁₇H₃₂SSn
MDL Number: MFCD09025807
CAS Number: 166766-89-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR