8-(Tributylstannyl)quinoline ≥95%

Supplier: Matrix Scientific

069556-500MG 069556-1G 069556-5G
101940-784EA 349.23 USD
101940-784 101940-786 101940-788
8-(Tributylstannyl)quinoline ≥95%
8-(Tributylstannyl)quinoline

Hazard: Irritant

Formula: C₂₁H₃₃NSn
MDL Number: MFCD09265482
CAS Number: 478282-21-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR